Learn More
Cellvis 6 Well glass bottom plate with high performance #1.5 cover glass, 20 Pack
Catalog No. NC0452316
Encompass
6 well glass bottom plate with high performance #1.5 cover glass (0.170±0.005mm). Black frame, with lid. Individually packed. Designed for high resolution imaging such as confocal microscopy. Suitable for long term tissue culture. Manufactured in a class 100,000 clean room. Plate made from virgin polystyrene, tissue culture treated. German high quality cover glass of superior optical quality, glass thickness is 0.170±0.005mmA USP class VI adhesive is used to assemble the cover glass and the plate. Sterilized by Gamma radiation. 20 per pack.
Catalog No. NC0452316 Supplier Cellvis Supplier No. P061.5HN
6 well glass bottom plate with high performance #1.5 cover glass (0.170±0.005mm). Black frame, with lid. Individually packed. Designed for high resolution imaging such as confocal microscopy. Suitable for long term tissue culture. Manufactured in a class 100,000 clean room. Plate made from virgin polystyrene, tissue culture treated. German high quality cover glass of superior optical quality, glass thickness is 0.170±0.005mmA USP class VI adhesive is used to assemble the cover glass and the plate. Sterilized by Gamma radiation. 20 per pack.