Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 6 well insert 1.0 um PET PK48 (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9301002
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 6-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for fast cell growth / barrier formation.
Catalog No. 50-267-3523