Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 6 well insert 5.0 um PET PK48 (cellQART)
Catalog No. 502673517
Encompass_Preferred
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 6-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for cell invasion, cell motility and chemotaxis.
Catalog No. 50-267-3517 Supplier Altenloh Brinck & Co US INC. Supplier No. 9305002
These cellQART Cell Culture Inserts come with PET membranes in translucent optics in 6-Well format. The Inserts are tissue culture treated, sterile and are compatible with most standard Cell Culture Plates. They are ideal for cell invasion, cell motility and chemotaxis.