Learn More
Thomson Instrument Company 6-Well Plate, 82mL, Square Well, Pyramid Bottom, Indiv. Wrapped w/Lid, Sterile
SDP
Catalog No. NC2950621
Encompass_Preferred
Thomson’s 6-Well Plates provide mid-scale working volumes (20–50 mL per well) with engineered gas exchange and mixing for reliable growth—bridging microscale screens and flask runs. Compatible with the Optimum Growth Integrated Lid and validated for stable operation on common shaker platforms.
•20–50 mL per well — flexible scale-up/scale-down for development studies.
•Enhanced gas exchange — promotes oxygenation for robust cell growth.
•Consistent mixing — uniform nutrient flow supports reproducible yields across wells.
•Environmental control — integrated lid reduces contamination and evaporation.
•High throughput — accommodates 24 plates per tray with broad shaker compatibility; validated with Infors™ Peg Tray Adapter and Kuhner™ Spring Tray F for secure, high-RPM shaking.
•User-friendly — replaces spin-tube workflows and avoids wrist strain from repetitive uncapping.
• Made in the USA
Catalog No. NC2950621 Supplier Thomson Instrument Company Supplier No. 931167
Thomson’s 6-Well Plates provide mid-scale working volumes (20–50 mL per well) with engineered gas exchange and mixing for reliable growth—bridging microscale screens and flask runs. Compatible with the Optimum Growth Integrated Lid and validated for stable operation on common shaker platforms.
•20–50 mL per well — flexible scale-up/scale-down for development studies.
•Enhanced gas exchange — promotes oxygenation for robust cell growth.
•Consistent mixing — uniform nutrient flow supports reproducible yields across wells.
•Environmental control — integrated lid reduces contamination and evaporation.
•High throughput — accommodates 24 plates per tray with broad shaker compatibility; validated with Infors™ Peg Tray Adapter and Kuhner™ Spring Tray F for secure, high-RPM shaking.
•User-friendly — replaces spin-tube workflows and avoids wrist strain from repetitive uncapping.
• Made in the USA