Learn More
Thomson Instrument Company 6-Well Plate, 82mL, Square Well, Pyramid Bottom, w/Integrated Lid, Sterile
SDP
Catalog No. NC3833616
Encompass_Preferred
The Thomson Optimum Growth 6-Well Plate with Integrated Lid (p/n 931167 + 981648) is a sterile, individually wrapped cultureware combo designed to support mid-scale mammalian and insect cell workflows. The square-well, pyramid-bottom plate enables uniform nutrient distribution and supports 20–50mL working volumes, validated with CHO and Sf21 cells for consistent growth and protein expression. The vented integrated lid ensures optimal gas exchange, minimizes evaporation, and reduces contamination risk, all while simplifying handling.
•20–50 mL working volume per well for development and scale-down studies.
•Uniform aeration & nutrient flow supports consistent growth and productivity.
•Platform-ready — validated with Infors™ Peg Tray Adapter and Kuhner™ Spring Tray F for secure, high-throughput shaking.
•Proven performance — studies show consistent titers across volumes and yields comparable to 50 mL spin tubes.
• Made in the USA
Catalog No. NC3833616 Supplier Thomson Instrument Company Supplier No. 931170
Add to Cart
Add to Cart
The Thomson Optimum Growth 6-Well Plate with Integrated Lid (p/n 931167 + 981648) is a sterile, individually wrapped cultureware combo designed to support mid-scale mammalian and insect cell workflows. The square-well, pyramid-bottom plate enables uniform nutrient distribution and supports 20–50mL working volumes, validated with CHO and Sf21 cells for consistent growth and protein expression. The vented integrated lid ensures optimal gas exchange, minimizes evaporation, and reduces contamination risk, all while simplifying handling.
•20–50 mL working volume per well for development and scale-down studies.
•Uniform aeration & nutrient flow supports consistent growth and productivity.
•Platform-ready — validated with Infors™ Peg Tray Adapter and Kuhner™ Spring Tray F for secure, high-throughput shaking.
•Proven performance — studies show consistent titers across volumes and yields comparable to 50 mL spin tubes.
• Made in the USA