Learn More
IMEB, Inc IMEB Slide Rack Staining Set With Holder, Plastic, 20 Slide, 6 Wells
SDP
Catalog No. NC9885647
Encompass
For use in histology, cytological or dermatology staining. Solution wells are deep enough to permit total immersion of 3"x1" slides. The plastic frame is solvent resistant with snap locking covers that permit complete portability, even when the wells are full. Dishes are placed in rack far enough from each other to preclude drippage of stain into other wells. Four white polypropylene and two green polypropylene dishes with tight fitting lids for each container, help to prevent evaporation of stain and solvents. Also available for purchase is a twelve solution wells set.
Catalog No. NC9885647 Supplier IMEB, Inc Supplier No. SSS60
For use in histology, cytological or dermatology staining. Solution wells are deep enough to permit total immersion of 3"x1" slides. The plastic frame is solvent resistant with snap locking covers that permit complete portability, even when the wells are full. Dishes are placed in rack far enough from each other to preclude drippage of stain into other wells. Four white polypropylene and two green polypropylene dishes with tight fitting lids for each container, help to prevent evaporation of stain and solvents. Also available for purchase is a twelve solution wells set.