Learn More
Bellco Glass, Inc. 6 Well Tissue Culture Plate
SDP
Catalog No. 502329833
Encompass_Preferred
CELLTREAT Glass bottom plates have the same great features as standard tissue culture plates with the added benefit of borosilicate glass bottom microwells. Convenient live cell viewing without needing to transfer. Optically clear borosilicate glass for exceptional viewing.Specifications Applications include cell culture and imaging live cell observation and specialty microscopy such as confocal high resolution and contrast Traditional footprints cause minimal disruption in cell culture techniques Constructed with noncytotoxic medical adhesive Round cover glass also available for use with glass bottom plates Case Quantity 5
Catalog No. 50-232-9833 Supplier Bellco Glass, Inc. Supplier No. 229107000
CELLTREAT Glass bottom plates have the same great features as standard tissue culture plates with the added benefit of borosilicate glass bottom microwells. Convenient live cell viewing without needing to transfer. Optically clear borosilicate glass for exceptional viewing.Specifications Applications include cell culture and imaging live cell observation and specialty microscopy such as confocal high resolution and contrast Traditional footprints cause minimal disruption in cell culture techniques Constructed with noncytotoxic medical adhesive Round cover glass also available for use with glass bottom plates Case Quantity 5