Learn More
Bellco Glass, Inc. 6 Well Tissue Culture Plate
Supplier: Bellco Glass, Inc. 229105000
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividualCase Quantity50SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area9.60cm²RNase/DNase FreeYes