Learn More
Chemglass Life Sciences 6in Diameter X 1/8in Thick Borosilicate Disc
SDP
Catalog No. CG190621
Encompass_Preferred
Plate glass, disc, borosilicate, 6in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.
Catalog No. CG190621 Supplier Chemglass Life Sciences Supplier No. CG190621
Plate glass, disc, borosilicate, 6in Diameter X 1/8in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.