Learn More
Labelmaster Inc Hazardous Waste Label, PVC-Free Film Stock
SDP
Catalog No. NC9693585
Encompass_Preferred
Hazardous Waste Accumulation Labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34. PVC-free film hazardous waste labels feature automotive-grade ink pigments.
Catalog No. NC9693585 Supplier Labelmaster Inc Supplier No. HWAV
Hazardous Waste Accumulation Labels provide a convenient way for a generator to mark accumulation containers in accordance with EPA regulations in 40 CFR part 262.34. PVC-free film hazardous waste labels feature automotive-grade ink pigments.