Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 6mm ID X 8mm OD
SDP
Catalog No. CGQ031005
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 6mm ID X 8mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031005 Supplier Chemglass Life Sciences Supplier No. CGQ031005
Graded Seal, Quartz to Borosilicate, 6mm ID X 8mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.