Learn More
Alkali Scientific 6 y y [Dimethylallylamino] Purine, 5 G
SDP
Catalog No. 5000661759
Encompass_Preferred
Dimethylallylamino purine (6-(dimethylallylamino) purine) is a cytokinin used to stimulate cell division and shoot proliferation in plant tissue culture. Available in 5 g amounts, this plant growth regulator is widely used to promote the regeneration of plants from tissue explants. Dimethylallylamino purine is essential in in vitro culture systems, particularly for enhancing shoot induction and promoting growth in a variety of plant species. It is a critical tool in plant biotechnology and genetic research, enabling the efficient propagation of plants, including genetically modified varieties. The 5 g package size is suitable for medium to large-scale laboratory applications and research projects.
Catalog No. 50-006-61759 Supplier Alkali Scientific Supplier No. YYD002CP
Dimethylallylamino purine (6-(dimethylallylamino) purine) is a cytokinin used to stimulate cell division and shoot proliferation in plant tissue culture. Available in 5 g amounts, this plant growth regulator is widely used to promote the regeneration of plants from tissue explants. Dimethylallylamino purine is essential in in vitro culture systems, particularly for enhancing shoot induction and promoting growth in a variety of plant species. It is a critical tool in plant biotechnology and genetic research, enabling the efficient propagation of plants, including genetically modified varieties. The 5 g package size is suitable for medium to large-scale laboratory applications and research projects.