Learn More
Alkali Scientific 6 y y [Dimethylallylamino] Purine Riboside, 100 Milligrams
SDP
Catalog No. 5000660523
Encompass_Preferred
Dimethylallylamino purine riboside is a cytokinin used in plant tissue culture to promote growth and regeneration of plant tissues. Available in 100 mg amounts, this reagent is commonly used in laboratory experiments aimed at improving the efficiency of plant regeneration. As a riboside form of dimethylallylamino purine, it is effective in stimulating cell division and shoot proliferation, making it invaluable in studies of plant growth and genetic modification. This reagent is often used in the development of plant biotechnology techniques, particularly those involving tissue culture and plant propagation. It plays a key role in increasing the success rate of regeneration protocols for various plant species.
Catalog No. 50-006-60523 Supplier Alkali Scientific Supplier No. YDI001CP
Dimethylallylamino purine riboside is a cytokinin used in plant tissue culture to promote growth and regeneration of plant tissues. Available in 100 mg amounts, this reagent is commonly used in laboratory experiments aimed at improving the efficiency of plant regeneration. As a riboside form of dimethylallylamino purine, it is effective in stimulating cell division and shoot proliferation, making it invaluable in studies of plant growth and genetic modification. This reagent is often used in the development of plant biotechnology techniques, particularly those involving tissue culture and plant propagation. It plays a key role in increasing the success rate of regeneration protocols for various plant species.