Learn More
Cayman Chemical 6 Z 9 Z 12 Z 15 Z 18 Z 21 ZTe
SDP
Supplier:  Cayman Chemical 1000516550 UG
Encompass_Preferred
A VLCPUFA; has been found in tunny, herring, cod-liver, pilot whale, sardine, and squalus-suclii-liver oils, and bovine retina; a component of triglycerides and cholesterol esters in mouse and rat testis; produced from dietary linolenic acid via a series of elongation and Δ6-desaturation reactions in fish and rat brain; undergoes β-oxidation to form DHA
Catalog No. 50-288-7959