Learn More
Cayman Chemical 6 Z 9 Z 12 Z 15 Z 18 Z 21 ZTe
SDP
Catalog No. 502890732
Encompass_Preferred
A VLCPUFA; has been found in tunny, herring, cod-liver, pilot whale, sardine, and squalus-suclii-liver oils, and bovine retina; a component of triglycerides and cholesterol esters in mouse and rat testis; produced from dietary linolenic acid via a series of elongation and Δ6-desaturation reactions in fish and rat brain; undergoes β-oxidation to form DHA
Catalog No. 50-289-0732 Supplier Cayman Chemical Supplier No. 1000516525 UG
A VLCPUFA; has been found in tunny, herring, cod-liver, pilot whale, sardine, and squalus-suclii-liver oils, and bovine retina; a component of triglycerides and cholesterol esters in mouse and rat testis; produced from dietary linolenic acid via a series of elongation and Δ6-desaturation reactions in fish and rat brain; undergoes β-oxidation to form DHA