Learn More
MOLTOX Molecular Toxicology Inc 60-250.5 - NADPH REGENSYS A Plus. 50ml refers to final volume of S9 mix/pkg. Intended for use in EAT testing. Supports 5-30% S9 mix. G-6-P MgCl2-KCL in 0.1M phosphate buffer. pH 7.4. Add to B Plus. Refrigerate.
SDP
Catalog No. NC3235400
Encompass
60-250.5 - NADPH REGENSYS A Plus. 50ml refers to final volume of S9 mix/pkg. Intended for use in EAT testing. Supports 5-30% S9 mix. G-6-P MgCl2-KCL in 0.1M phosphate buffer. pH 7.4. Add to B Plus. Refrigerate.
Catalog No. NC3235400 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 60250.5
Add to Cart
Add to Cart
60-250.5 - NADPH REGENSYS A Plus. 50ml refers to final volume of S9 mix/pkg. Intended for use in EAT testing. Supports 5-30% S9 mix. G-6-P MgCl2-KCL in 0.1M phosphate buffer. pH 7.4. Add to B Plus. Refrigerate.