Learn More
Chemglass Life Sciences 60/50 Outer Joint, Full Length, on 64mm OD x 2.4mm W tubing
SDP
Supplier:  Chemglass Life Sciences CG10113
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 60/50 Outer Joint, Full Length, on 64mm OD x 2.4mm W tubing.
Catalog No. CG10113