Learn More
Terumo™ 60 cc Syringes
Catalog No. 22290022 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Connection Type:
Catheter
Eccentric Luer Slip
Luer Lock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
22-290-022 Luer Lock
22-290-023 Eccentric Luer Slip
22-290-024 Catheter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-290-022 Supplier Terumo™ Supplier No. SS60L
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
60 cc Syringes are available with various style tips to suit application.
  • Superior sharpness and smoothness
  • Ultra Sharp Needles
  • Clear Barrels
  • Bold, Precise Scale Markings
  • Smooth Plunger Action
  • Tamper Evident Peel Packs
Specifications
Material Plastic
Color Clear
Includes Syringe Only
Latex Free Yes
Product Type Syringe
Disposable Single-use
Graduations 1 cc
Quantity 25 Pk., 4 Pk./Cs.
Volume (Metric) 60 mL
Connection Type Luer Lock
Graduated Yes
Sterility Sterile