Learn More
Justrite™ 60 Gallon Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet
GREENER_CHOICE
Catalog No. 19822396 Shop All Justrite Safety Group Products
Change view
Click to view available options
Door Style:
Self-close
Self-close, Bi-fold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
19-822-396 Self-close
19-822-388 Self-close, Bi-fold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-396 Supplier Justrite™ Supplier No. 896022
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 60 Gallon Sure-Grip™ EX Corrosives/Acid Steel Safety Cabinet is specially designed for safely storing acids and corrosives in laboratories.
Justrite blue steel safety cabinets for corrosives have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet. Self-close door(s) shuts and latches automatically when a fusible link melts at 165°F (74°C) under fire conditions. Unique, concealed self-closing mechanism offers obstruction-free access to contents.
Specifications
Load Bearing Capacity 60 gal.
Description Cold-rolled Steel (18 ga)
Dimensions (D x W x H) 34 x 34 x 65 in.
Door Style Self-close
Material Steel (CR)
Color Blue