Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ 60 Gallon Yellow Infectious Linen Bag

Catalog No. 17988504 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
60 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-988-504 60 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-988-504 Supplier Eagle™ Supplier No. 1694
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yellow infectious linen bag used to minimize waste by segregating infectious linen to be laundered

  • Designed to fit Eagle™ 1655 (nestable) and 1656 (straight sided) series drums
  • Once bag is full and ready for laundering, seal bag in place and close drum using its lid and lever-lock
  • Yellow color bag is easily identifiable as infectious linen
  • Drums can be yellow, black, or blue
  • Printing includes English, Spanish and French translation along with international recognizable biohazard symbol
  • Material: linear low density polyethylene (LLDPE)
  • Thickness: 1.2 mil
  • High level of flexibility and shear resistance can be attained without leaks
  • Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
  • Tubular construction, single side seal and star seal bottom provide stable, leak proof bag
  • 100/Pk.

Specifications

Capacity (English) 60 gal.
Color Yellow
Quantity 100/Pk.
Autoclavable Not Autoclavable
Liner Material LDPE
For Use With (Equipment) Eagle™ Drums
Width (English) 38 in.
Length (English) 58 in.
Thickness 1.2 mil
Height (English) 38 in.
Material Polyethylene
Product Type Biohazard Linen Bags
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.