Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somni Scientific™ 60 in. Bain Non-rebreathing Circuit
SDP

Catalog No. 17377522 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
Rabbit, Dog, Cat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
17-377-522 Rabbit, Dog, Cat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-377-522 Supplier Somni Scientific™ Supplier No. BC13005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This non-rebreathing CPRAM/Bain circuit tube within a tube design provides heat and moisture conservation with fresh gas entering the circuit at the patient end.

  • For use with SOMNI™ Bain non-rebreathing system
  • GS elbow included
  • For use with animals under 7.5 kg, such as rabbit, dog, or cat

Specifications

Autoclavable Not Autoclavable
Animal Rabbit, Dog, Cat
For Use With (Application) Surgical Procedures
Type Breathing Circuit
Disposable Reusable
For Use With (Equipment) Bain Non-rebreathing Circuit
Sterility Non-sterile
Length (English) 60 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.