Learn More
Chemrus Inc 60 ML LIU FLASK 8/PACK
Catalog No. NC1444286
Encompass
LIU FLASK, FLAT BOTTOM, BARREL-SHAPE, HEAVY WALL, SINGLE NECK, 60ml, 24/40 taper outer joint, 8/pack
Catalog No. NC1444286 Supplier Chemrus Inc Supplier No. CR240160
LIU FLASK, FLAT BOTTOM, BARREL-SHAPE, HEAVY WALL, SINGLE NECK, 60ml, 24/40 taper outer joint, 8/pack