Learn More
Savillex Corporation Savillex 60 mL Standard Jar, High Chemical Resistance, Seamless Design, PFA Material, 60 mL
SDP
Catalog No. NC1921370
Encompass
The Savillex 60 mL Standard Jar is an essential laboratory container designed to securely store and handle various materials. Its seamless, translucent PFA construction ensures superior chemical resistance and visibility. Enhanced with a wide-mouth design and ribbed areas for easy handling, this jar supports safe storage across a wide temperature range. Ideal for critical scientific applications, it offers unparalleled purity and safety features that differentiate it from standard jars.
Catalog No. NC1921370 Supplier Savillex Corporation Supplier No. 100006001
The Savillex 60 mL Standard Jar is an essential laboratory container designed to securely store and handle various materials. Its seamless, translucent PFA construction ensures superior chemical resistance and visibility. Enhanced with a wide-mouth design and ribbed areas for easy handling, this jar supports safe storage across a wide temperature range. Ideal for critical scientific applications, it offers unparalleled purity and safety features that differentiate it from standard jars.