Learn More
Electron Microscopy Sciences Syringe With Adapter 60 ml
SDP
Supplier:  Electron Microscopy Sciences 72486
Encompass_Preferred
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 60 ml syringe dispenses 1000 to 5000 uL.
Type: Not autoclavable
Catalog No. 50-949-641