Learn More
Electron Microscopy Sciences BD* Carbon Steel Scalpel Blade #60
SDP
Catalog No. 50949734
Encompass_Preferred
Made from surgical carbon steel, size 60 with convex cutting edge. The most useful in histology labs. These scalpel blades are 2 3/8" (6cm) long and ribbed for extra rigidity. They are non-sterile and are wrapped individually. Bard-Parker brand.
Catalog No. 50-949-734 Supplier Electron Microscopy Sciences Supplier No. 7205010
Made from surgical carbon steel, size 60 with convex cutting edge. The most useful in histology labs. These scalpel blades are 2 3/8" (6cm) long and ribbed for extra rigidity. They are non-sterile and are wrapped individually. Bard-Parker brand.