Learn More
American Diagnostic Corporation 60 SECOND DIGITAL THRMTR 20/CS
SDP
Catalog No. NC1864976
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
The Adtemp 412 features:Super low-cost digital thermometerCompact size: less than 5 long, weighs just 14 gramsRange 90.0F to 109.9F .2F or 32.0C to 43.9C .1C depending upon scale selectionNon-replaceable 1.55V (LR41) type battery provides up to 4,500 measurementsIntegral storage case keeps instrument sanitaryBulk unit packaged 20/box, 240/caseMeets the accuracy requirements for intermittent thermometers under ASTM E1112This product not available for purchase in Canada.
This item is not returnable. View return policy
Catalog No. NC1864976 Supplier American Diagnostic Corporation Supplier No. 41200
This item has a minimum qty of 2 per supplier requirements.
The Adtemp 412 features:Super low-cost digital thermometerCompact size: less than 5 long, weighs just 14 gramsRange 90.0F to 109.9F .2F or 32.0C to 43.9C .1C depending upon scale selectionNon-replaceable 1.55V (LR41) type battery provides up to 4,500 measurementsIntegral storage case keeps instrument sanitaryBulk unit packaged 20/box, 240/caseMeets the accuracy requirements for intermittent thermometers under ASTM E1112This product not available for purchase in Canada.