Learn More
Bellco Glass, Inc. 60 x 12 mm Petri Dish, sterile 20/bag, 500/cs
SDP
Catalog No. 502330246
Encompass_Preferred
60 x 12 mm Petri Dish, sterile 20/pk, 500/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic
Catalog No. 50-233-0246 Supplier Bellco Glass, Inc. Supplier No. 451400475
60 x 12 mm Petri Dish, sterile 20/pk, 500/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic