Learn More
Uline Standard Mobile Stainless Steel Worktable with Bottom Shelf - 60 x 30, Stainless Steel
Catalog No. NC2132592
Encompass
Easy portability for reconfiguring and cleaning work area, Galvanized legs and shelf Type 430 top, 1 5/8 diameter steel tube legs 35 table height, Adjustable bottom shelf, 4 polyurethane locking swivel casters, NSF certified, Optional Overshelves and Drawer available
This item is not returnable. View return policy
Catalog No. NC2132592 Supplier Uline Supplier No. H9646
Easy portability for reconfiguring and cleaning work area, Galvanized legs and shelf Type 430 top, 1 5/8 diameter steel tube legs 35 table height, Adjustable bottom shelf, 4 polyurethane locking swivel casters, NSF certified, Optional Overshelves and Drawer available