Learn More
Zymo Research Corporation 600 ml Reservoir
SDP
Catalog No. 502037006
Encompass_Preferred
The 600 ml Reservoir, used in conjunction with a luer-lock column, can be used with vacuum manifolds for the purification of DNA and/or RNA. The large volume capacity is perfect for large-scale purification such as plasmid Gigapreps (e.g. ZymoPURE Gigaprep). The reservoir is designed to ensure no liquid is left behind during processing. The reservoir features durable polypropylene construction. The volume capacity of the reservoir is 600 ml.
Catalog No. 50-203-7006 Supplier Zymo Research Corporation Supplier No. C10335
The 600 ml Reservoir, used in conjunction with a luer-lock column, can be used with vacuum manifolds for the purification of DNA and/or RNA. The large volume capacity is perfect for large-scale purification such as plasmid Gigapreps (e.g. ZymoPURE Gigaprep). The reservoir is designed to ensure no liquid is left behind during processing. The reservoir features durable polypropylene construction. The volume capacity of the reservoir is 600 ml.