Learn More
Stoelting™ 600 Series Syringe
SDP
Catalog No. 10000918 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Blunt tip
Catalog No. Model
10-000-918 Blunt tip
1 options
Catalog No. 10-000-918 Supplier Stoelting™ Supplier No. 53491
Add to Cart
Add to Cart
Stoelting™ 600 Series Syringe is a half stroke version of the original 700 series syringes.
  • Ideal for dispensing volumes from 0.25 μL up to 5 μL
  • Often used in applications that require one handed operation because of the short stroke length and the reinforced plunger
Specifications
For Use With (Application) Neuroscience
Size 2.5 μL; 22 gauge
Model Blunt tip