Learn More
Glass Expansion Nickel Skimmer Cone for PerkinElmer Elan 6000/9000/DRCII/DRC-E (WE021137)
SDP
Catalog No. NC1585740
Encompass_Preferred
Nickel Skimmer Cone (WE021137) ¿ Standard skimmer cone for PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Replace skimmer cone O-ring if brittle, cracked or damaged (NC1585742). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1022.For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number 501907683.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1585740 Supplier Glass Expansion Supplier No. PE2012NI
Nickel Skimmer Cone (WE021137) ¿ Standard skimmer cone for PerkinElmer Elan 6000/9000/DRCII/DRC-E ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Replace skimmer cone O-ring if brittle, cracked or damaged (NC1585742). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection. Use skimmer ConeGuard to protect cone threads during cleaning, 70-803-1022.For corrosive samples, including HF, we recommend Platinum Skimmer Cones, see catalog number 501907683.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001