Learn More
3M™ 6000 Series Probed Reusable Respirator Assembly
Catalog No. 179873B Shop All 3M Products
Change view
Click to view available options
Model:
6200 Q
6300 Q
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
17-987-3B 6300 Q Large
17-987-3A 6200 Q Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-987-3B Supplier 3M™ Supplier No. 7000126031
Add to Cart
Add to Cart
This item is not returnable. View return policy
Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.
Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing
  • Probed facepiece allows for numerical measurement of the amount of leakage into the respirator and includes a probed facepiece and P100 filters
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Quantitative Fit Test Assembly
Certifications/Compliance EN 140:1998, NIOSH 42 CFR Part 84
For Use With (Application) Conduct Quantitative Fit Tests on Respirator 6000 Series
For Use With (Equipment) Conduct Quantitative Fit Tests on Respirator 6000 Series
Includes Probed facepiece and P100 filter
Material Thermoplastic Elastomer
Model 6300 Q
Size Large