Learn More
Sino Biological Recombinant Ferret CD8 alpha Protein (His Tag)
Supplier:  Sino Biological 60001F08H100
Encompass_Preferred
A DNA sequence encoding the Ferret (Mustela putorius furo) CD8A (ABS50091.1) extracellular domain (Met 1-Glu 186) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-3207