Learn More
Sino Biological Recombinant Ferret CD20 Protein (His Tag)
Supplier:  Sino Biological 60004F08H100
Encompass_Preferred
A DNA sequence encoding the C-terminal cytoplasmic domain of Ferret (Mustela putorius furo) CD20 (Glu 213-Pro 297) was expressed, with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-162-3214