Learn More
Sino Biological Recombinant Ferret IFN gamma Protein (His Tag)
Supplier:  Sino Biological 60007F08H100
Encompass_Preferred
A DNA sequence encoding the ferret IFNG (A4PIZ9) (Met1-Lys166) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3216