Learn More
Ace Glass, Inc. 600mL jacketed filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 83mm IDx185mm depth, with CAPFE O-ring, #11 Ace-Thred inlet/outlet connections, use with 3/8in ID tubing, pressure rated at 35 psi.
SDP
Catalog No. 50972809
Encompass
600mL jacketed filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 83mm IDx185mm depth, with CAPFE O-ring, #11 Ace-Thred inlet/outlet connections, use with 3/8in ID tubing, pressure rated at 35 psi.
Catalog No. 50-972-809 Supplier Ace Glass, Inc. Supplier No. 6386312
600mL jacketed filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 83mm IDx185mm depth, with CAPFE O-ring, #11 Ace-Thred inlet/outlet connections, use with 3/8in ID tubing, pressure rated at 35 psi.