Learn More
Ace Glass, Inc. Beaker, 600mL, jacketed, Ace-Safe hose connections for 3/8intubing, 152mm inside height, 81mm inside diameter
SDP
Catalog No. 50978356
Encompass_Preferred
600mL jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 152mm I.H. x 81mm I.D. Does not have a pouring spout.
Catalog No. 50-978-356 Supplier Ace Glass, Inc. Supplier No. 5340115
600mL jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 152mm I.H. x 81mm I.D. Does not have a pouring spout.