Learn More
Ace Glass, Inc. 600mL filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 83mm IDx185mm depth, with CAPFE O-ring, pressure rated at 35 psi.
SDP
Catalog No. 50972788
Encompass
Catalog No. 50-972-788 Supplier Ace Glass, Inc. Supplier No. 638411