Learn More
Chemglass Life Sciences 600mL Filter Funnel, Buchner, 24/40 Lower Vacuum Assembly, Fine Frit
SDP
Catalog No. CG140632
Encompass_Preferred
600mL Filter Funnel, Buchner, 24/40 Lower Vacuum Assembly, Fine Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.
Catalog No. CG140632 Supplier Chemglass Life Sciences Supplier No. CG140632
600mL Filter Funnel, Buchner, 24/40 Lower Vacuum Assembly, Fine Frit. Buchner style filter funnel having a lower standard taper inner joint and serrated tubulation for connection to a vacuum source. Disc diameters and height above the disc are the same as CG-1402.