Learn More
Ace Glass, Inc. Flask, freeze drying, 600mL, 282mm height, 53mm ID neck, 60mm body OD, includes snap-on rubber cap and 1/2in ODPTFE plain manifold connection adapter
SDP
Catalog No. 5097423
Encompass_Preferred
Catalog No. 50-974-23 Supplier Ace Glass, Inc. Supplier No. 703509