Learn More
Bellco Glass, Inc. 600mL Graduated Beaker
SDP
Catalog No. 502329602
Encompass_Preferred
The Polypropylene graduated beaker has molded  pouring spouts and rolled up lip for dripless work environment. They have angle walls for easier handling and sample retrieving. Nesting design for easy stacking and storage.Specifications Smaller case quantities are easier to store and are ideal for small scale users. Polypropylene material fir resistance to a variety of strong acids and solvents. Size 600mL Pack Description 25/Bag Case Quantity 25
Catalog No. 50-232-9602 Supplier Bellco Glass, Inc. Supplier No. 230517000
The Polypropylene graduated beaker has molded  pouring spouts and rolled up lip for dripless work environment. They have angle walls for easier handling and sample retrieving. Nesting design for easy stacking and storage.Specifications Smaller case quantities are easier to store and are ideal for small scale users. Polypropylene material fir resistance to a variety of strong acids and solvents. Size 600mL Pack Description 25/Bag Case Quantity 25