Learn More
Ace Glass, Inc. 600mL Instatherm filter reactor body with 60mm ID grooved top flange, #50 bottom thread, 83mm IDx185mm depth, with CAPFE O-ring and power cord, 40v/8amp max
SDP
Catalog No. 50972820
Encompass
600mL Instatherm coated filter reactor body with a 60mm I.D. grooved top flange for connection to a reaction head with the use of a quick release clamp, and a #50 Ace-Thred bottom thread for use with a bottom adapter with a filter support and drain valve. Instatherm provides safer, rapid heating, and can be operated at 115v with ACE Proportional Temperature Controllers, or with a Powerstat. Reactor body supplied with a CAPFE o-ring and a power cord. Dimensions: 83mm I.D. x 185mm D.
Catalog No. 50-972-820 Supplier Ace Glass, Inc. Supplier No. 638820
600mL Instatherm coated filter reactor body with a 60mm I.D. grooved top flange for connection to a reaction head with the use of a quick release clamp, and a #50 Ace-Thred bottom thread for use with a bottom adapter with a filter support and drain valve. Instatherm provides safer, rapid heating, and can be operated at 115v with ACE Proportional Temperature Controllers, or with a Powerstat. Reactor body supplied with a CAPFE o-ring and a power cord. Dimensions: 83mm I.D. x 185mm D.