Learn More
Ace Glass, Inc. Rusek pressure filter/drying funnel, 600mL, porosity C(25-50 microns), includes 7855-746 FETFE; O-ring, use with 7669-20 clamp
SDP
Catalog No. 50974328
Encompass_Preferred
600mL porosity C (25-50 micron) Rusek pressure filter/drying funnel used for drying air-sensitive compounds with inert gas, or a simple pressure filter funnel. O-ring joint connects the cap to the body to give a larger opening. Rusek funnel will withstand pressures of 20psi as supplied, but NO warranty, expressed or implied. Cap adapter has serrated hose connection for pressure source, usually a rubber bulb. O-ring joint on cap matches the funnel body joint, and secures with a pinch clamp. Includes a FETFE o-ring. Pinch clamp (Item 509557983) and rubber bulb sold separately.
Catalog No. 50-974-328 Supplier Ace Glass, Inc. Supplier No. 719055
Add to Cart
Add to Cart
600mL porosity C (25-50 micron) Rusek pressure filter/drying funnel used for drying air-sensitive compounds with inert gas, or a simple pressure filter funnel. O-ring joint connects the cap to the body to give a larger opening. Rusek funnel will withstand pressures of 20psi as supplied, but NO warranty, expressed or implied. Cap adapter has serrated hose connection for pressure source, usually a rubber bulb. O-ring joint on cap matches the funnel body joint, and secures with a pinch clamp. Includes a FETFE o-ring. Pinch clamp (Item 509557983) and rubber bulb sold separately.