Learn More
Ace Glass, Inc. 600mL Buchner filter funnel with porosity E (4-8 micron) ACE fritted glass disc.
SDP
Supplier:  Ace Glass, Inc. 718670
Encompass_Preferred
600mL Buchner filter funnel with porosity E (4-8 micron) ACE fritted glass disc.
Catalog No. 50-974-289