Learn More
Ace Glass, Inc. 600ML VESSEL 600ML VESSEL
SDP
Catalog No. 50978848
Encompass_Preferred
600mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred, for connection to an umbrella manifold. Vessel has sloped sides. Working capacity of vessel is about 1/2 of volume.
Catalog No. 50-978-848 Supplier Ace Glass, Inc. Supplier No. 669640
600mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred, for connection to an umbrella manifold. Vessel has sloped sides. Working capacity of vessel is about 1/2 of volume.