Learn More
Hach Company ORP reference solution, 600 mV, 500 mL
Catalog No. 501997409 Shop All Hach Company Products
Encompass
ORP Reference Solution used for ORP sensor calibration (in resealable plastic bottles).
Catalog No. 50-199-7409 Supplier Hach Company Supplier No. 25M2A1002115
ORP Reference Solution used for ORP sensor calibration (in resealable plastic bottles).