Learn More
MSC CM Mini-Ratchet Hoist, 1100 lb, 5 ft, 1 ea
Catalog No. 501299363
Encompass
The CM Mini-Ratchet lever hoist is a lightweight, compact, and portable tool designed for pulling, lifting, and tensioning applications in confined spaces. It features a plated load chain, Weston-type load brake, and a rubber grip handle for secure, one-hand operation.
  • Lightweight and compact design
  • Through-handle design for reduced headroom
  • Operates in any position
  • Aluminum frame and zinc-plated steel parts for corrosion resistance
  • Self-storing, hardened steel chain
  • Impact resistant, all-steel frame, gear case, and cover
  • Forged upper and lower hooks with latches
  • Hooks open slowly when overloaded for safety
  • Free chaining feature for quick load attachment
  • Tested to 125% of rated capacity
Catalog No. 50-129-9363 Supplier MSC Supplier No. 59534834
The CM Mini-Ratchet lever hoist is a lightweight, compact, and portable tool designed for pulling, lifting, and tensioning applications in confined spaces. It features a plated load chain, Weston-type load brake, and a rubber grip handle for secure, one-hand operation.
  • Lightweight and compact design
  • Through-handle design for reduced headroom
  • Operates in any position
  • Aluminum frame and zinc-plated steel parts for corrosion resistance
  • Self-storing, hardened steel chain
  • Impact resistant, all-steel frame, gear case, and cover
  • Forged upper and lower hooks with latches
  • Hooks open slowly when overloaded for safety
  • Free chaining feature for quick load attachment
  • Tested to 125% of rated capacity