Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ 6051020 Pro Series ISE/DO Cable

Catalog No. 1339892 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Length (English):
39.3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
13-398-92 39.3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-398-92 Supplier YSI™ Supplier No. 60510201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cable for Pro Plus DO/pH/temperature meters

  • Measure any ISE (pH, ORP, pH/ORP combo, ammonium, nitrate, chloride), dissolved oxygen and temperature
  • Cables over 1-meter also come with the 603062 cable management kit
  • The 6051020 cables ONLY work with the YSI Pro Plus handheld
  • DOES NOT come with the ISE sensor

Order Info

Order probes separately.

TRUSTED_SUSTAINABILITY

Specifications

Length (English) 39.3 in.
Length (Metric) 1 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.