Learn More
eMolecules​ 6089-09-4 | 4-Pentynoic acid | Ambeed | MFCD00004407 | 98.101 | C5H6O2 | 98.000 | OC(=O)CCC#C | 1g | 552723089
Catalog No. 5000482843 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Pentynoic acid | Ambeed | 6089-09-4 | MFCD00004407 | 98.101 | C5H6O2 | 98.000 | OC(=O)CCC#C | 1g | 552723089
Catalog No. 50-004-82843 Supplier eMolecules​ Supplier No. 552723089
4-Pentynoic acid | Ambeed | 6089-09-4 | MFCD00004407 | 98.101 | C5H6O2 | 98.000 | OC(=O)CCC#C | 1g | 552723089