Learn More
Leica Biosystems 60B RND TIP SCALPL BLADE 100/C
Catalog No. NC9883927
Encompass_Preferred
A diverse range of scalpel blades and handles for use in the pathology laboratory and mortuary. #22 and #60scalpels are available in the traditional pointed tip format or in the #22B and #60B rounded tip or bullnose format for enhanced user safety. These scalpels are made of finest carbon steel and are non-sterile.
Catalog No. NC9883927 Supplier Leica Biosystems Supplier No. 3802187
A diverse range of scalpel blades and handles for use in the pathology laboratory and mortuary. #22 and #60scalpels are available in the traditional pointed tip format or in the #22B and #60B rounded tip or bullnose format for enhanced user safety. These scalpels are made of finest carbon steel and are non-sterile.